CRM8064
Analytes B Standard for CLP-M with ICP/MS
for ICP-MS, certified reference material, in nitric acid (2%, varied conc.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
Quality Level
packaging
bottle of 125 mL
concentration
in nitric acid (2%, varied conc.)
format
multi-component solution
storage temp.
room temp
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Analyte
Description
Ag 5 μg/mL
As 10 μg/mL
Cd 10 μg/mL
Co 20 μg/mL
Cr 20 μg/mL
Cu 20 μg/mL
Mn 20 μg/mL
Ni 20 μg/mL
V 20 μg/mL
Se 10 μg/mL
Zn 10 μg/mL
See All (11)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service