Skip to Content
Merck
All Photos(5)

Key Documents

CRM09001L

Millipore

Escherichia coli NCTC 9001 Lenticule® discs

Escherichia coli NCTC 9001 Lenticule® discs
1 of 1 reviewers received a sample product or took part in a promotion

30-120 CFU, suitable for microbiology, certified reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

E. coli NCTC 9001 Lenticule® discs, Escherichia coli NCTC 9001 Lenticule® discs, Certified reference material, Microbiological standards

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
CHF 85.60

CHF 85.60


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
10 EA
CHF 85.60

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CHF 85.60


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

certified reference material

Quality Level

Agency

ISO 17034
ISO/IEC 17025

form

disc (hydrosoluble)
solid

shelf life

limited shelf life, expiry date on the label

packaging

pack of 10

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

environmental
food and beverages

microbiology

storage temp.

−20°C

suitability

Escherichia coli
Escherichia spp.
coliforms

Looking for similar products? Visit Product Comparison Guide

General description

Escherichia coli NCTC 9001 Lenticule® discs are Certified Microorganisms Standards (Certified Microbiological Reference Materials), which can be used as quantitative and qualitative control organisms. They contain colony forming units (CFUs) in the range of 30-120 CFU per disc.

The technology allows long-term conservation and renders the organisms resistant to temperature changes. They are intended to be used as daily internal quality control organisms.

These microorganisms are certified according to ISO/IEC 17025 and produced under reproducible conditions compliant with ISO 17034 using authenticated strains from NCTC. These ready-to-use microbiological controls minimize the need to maintain control strains in the test laboratory and guarantee an authenticated control culture for every quality control test.

Lenticule® discs are prepared from a traceable culture obtained freeze-dried from the National Collection of Type Cultures (NCTC) or National Collection of Pathogenic Fungi (NCPF) and are manufactured by Sigma-Aldrich under license from UK Health Security Agency.

Application

Escherichia coli NCTC 9001 Lenticule® discs are used as reference strains in routine quality control for food, beverage, water, and environmental testing. Additionally, they are excellent for use in method development and validations, as starter cultures, and for use in research studies and educational purposes.

Features and Benefits

  • Ready-to-use at convenient CFU ranges
  • No dilution required
  • Quick rehydration in 10 minutes
  • At least 16-month shelf life in standard -20 °C freezer
  • Complete with comprehensive certificate of analysis
  • No maintenance of stock cultures
  • No recovery time or pre-enrichment step necessary
  • Robust, efficient, and reliable
  • Easy-to-use
  • Cost-effective

Other Notes

Strain origin: NCTC 9001

Other cell culture collections:

DSM 30083

ATCC® 11775

CECT 515

Click here to access the BacDive database for more info on metabolism, genome, growth media, and more
Strain History - Accessioned into the NCTC in 1952 < Dr F. Kauffmann (Statens Seruminstitut, Copenhagen, Denmark). Isolated from human urine (case of cystitis), geographic origin unknown, before 1953.

Legal Information

ATCC is a registered trademark of American Type Culture Collection
Lenticule is a registered trademark of The Secretary of State for Health (United Kingdom)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.

Questions

  1. Is this strain of E.coli is non pathogenic???

    1 answer
    1. The E. coli strain has a BSL 2 level for biosafety and is a pathogenic strain. Please access the BacDive database for additional information, such as metabolism, genome, and growth medium: https://bacdive.dsmz.de/strain/4907.

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service