Skip to Content
Merck
All Photos(2)

Documents

C4916

Sigma-Aldrich

5-Carboxyfluorescein diacetate

~95% purity (HPLC), powder

Synonym(s):

5-CFDA, 5-Carboxy-di-O-acetylfluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H16O9
CAS Number:
Molecular Weight:
460.39
Beilstein:
68265
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

5-Carboxyfluorescein diacetate, ~95% (HPLC)

Assay

~95% (HPLC)

Quality Level

form

powder

solubility

chloroform/ethanol (1:1): 9.80-10.20 mg/mL, clear

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

CC(=O)Oc1ccc2c(Oc3cc(OC(C)=O)ccc3C24OC(=O)c5cc(ccc45)C(O)=O)c1

InChI

1S/C25H16O9/c1-12(26)31-15-4-7-19-21(10-15)33-22-11-16(32-13(2)27)5-8-20(22)25(19)18-6-3-14(23(28)29)9-17(18)24(30)34-25/h3-11H,1-2H3,(H,28,29)

InChI key

WPUZGNPQMIWOHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Carboxyfluorescein diacetate has been used for the staining of bacteria to study bacterial adhesion to mucin.

Biochem/physiol Actions

5-Carboxyfluorescein diacetate is required for the staining of cells to differentiate between live and dead cells. It helps in monitoring integrity and intactness of cell membrane. 5-Carboxyfluorescein diacetate is also used for the measurement of intracellular pH. 5-Carboxyfluorescein diacetate is a non-fluorescent molecule which is cleaved by intracellular esterases, thereby giving a green fluorescent product carboxyfluorescein. In viable cells, the cellular membrane is impermeable to the fluorescent molecule.

comparable product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Wei Ouyang et al.
Cell transplantation, 13(6), 649-657 (2005-01-15)
Bone marrow stromal cells are potentially useful for tendon repair and regeneration. To provide lasting benefits, the seeded cells must survive implantation at local tendon sites. Our objective was to determine the in vivo fate of allogeneic bone marrow stromal
Maillard-type glycoconjugates from dairy proteins inhibit adhesion of Escherichia coli to mucin.
Food Chemistry, 129, 1435-1443 (2011)
Arend W Overeem et al.
Journal of hepatology, 71(2), 344-356 (2019-04-10)
Hepatocyte polarity is essential for the development of bile canaliculi and for safely transporting bile and waste products from the liver. Functional studies of autologous mutated proteins in the context of the polarized hepatocyte have been challenging because of the
Isabel Comino et al.
Food & nutrition research, 60, 30324-30324 (2016-02-09)
Oats provide important nutritional and pharmacological properties, although their safety in coeliac patients remains controversial. Previous studies have confirmed that the reactivity of the anti-33-mer monoclonal antibody with different oat varieties is proportional to the immune responses in terms of
Dietary glycosaminoglycans interfere in bacterial adhesion and gliadin-induced pro-inflammatory response in intestinal epithelial (Caco-2) cells.
Laparra JM, et al.
International Journal of Biological Macromolecules, 47, 458-464 (2010)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service