Skip to Content
Merck
All Photos(1)

Documents

C0299000

Calcium glucoheptonate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Calcium gluceptate, α-D-Glucoheptonic acid calcium salt, D-Glycero-D-glucoheptonic acid calcium salt, Calcium α-D-heptagluconate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26CaO16
CAS Number:
Molecular Weight:
490.42
Beilstein:
6104529
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

calcium glucoheptonate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Ca++].OC[C@H](O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)C([O-])=O.OC[C@H](O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)C([O-])=O

InChI

1S/2C7H14O8.Ca/c2*8-1-2(9)3(10)4(11)5(12)6(13)7(14)15;/h2*2-6,8-13H,1H2,(H,14,15);/q;;+2/p-2

InChI key

FATUQANACHZLRT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Calcium glucoheptonate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prashant Kumar Modi et al.
PloS one, 14(9), e0222240-e0222240 (2019-09-10)
Calcium is the key macromineral having a role in skeletal structure and function, muscle contraction, and neurotransmission. Bone remodeling is maintained through a constant balance between calcium resorption and deposition. Calcium deficiency is resolved through calcium supplementation, and among the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service