BP555
2-Trifluoromethylthioxanthone
British Pharmacopoeia (BP) Reference Standard
Synonym(s):
2-(Trifluoromethyl)thioxanthen-9-one
About This Item
Recommended Products
API family
flupentixol
form
solid
manufacturer/tradename
BP
mp
147-151 °C (lit.)
storage temp.
2-8°C
SMILES string
FC(F)(F)c1ccc2Sc3ccccc3C(=O)c2c1
InChI
1S/C14H7F3OS/c15-14(16,17)8-5-6-12-10(7-8)13(18)9-3-1-2-4-11(9)19-12/h1-7H
InChI key
NEWRXGDGZGIHIS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Also used in monographs such as:
Packaging
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service