Skip to Content
Merck
All Photos(1)

Key Documents

BCR651

Beer (EtOH, low level)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
CHF 92.80

CHF 92.80


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
10 ML
CHF 92.80

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

CHF 92.80


Estimated to ship on30 May 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

food and beverages

format

matrix material

storage temp.

2-8°C

General description

The material is related to the labeling of ′alcohol-free′ or ′de-alcoholised′ and ′low alcohol′ beverages according to Directive 90/496/EC.
Ethanol 0.505 ± 0.06 % ethanol v/v

Packaging

Amber glass ampule, flushed with nitrogen, containing 10 mL of sample.

Analysis Note

For more information please see:
BCR651

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Ethanol

Matrix Group: Beverages and Water

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service