Skip to Content
Merck
All Photos(1)

Key Documents

BCR290

2,3,3′-Trichlorobiphenyl

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CHF 345.00

CHF 345.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
CHF 345.00

About This Item

Empirical Formula (Hill Notation):
C12H7Cl3
CAS Number:
Molecular Weight:
257.54
Beilstein:
8141481
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

CHF 345.00


Estimated to ship on14 May 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

neat

storage temp.

2-8°C

SMILES string

Clc1cccc(c1)-c2cccc(Cl)c2Cl

InChI

1S/C12H7Cl3/c13-9-4-1-3-8(7-9)10-5-2-6-11(14)12(10)15/h1-7H

InChI key

SXHLTVKPNQVZGL-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR290

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of sediment contact and uptake mechanisms on accumulation of three chlorinated hydrocarbons in the midge, Chironomus riparius.
D M Fry et al.
Bulletin of environmental contamination and toxicology, 44(5), 790-797 (1990-05-01)
Hu Cheng et al.
Journal of chromatography. A, 1548, 1-9 (2018-03-21)
Porous materials have great prospective applications for solid-phase microextraction (SPME) technology because of their large specific surface area and pore volume. In this study, a hierarchical porous carbon (HPC) was synthesized by simple hydrothermal reaction and potassium hydroxide (KOH) activation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service