Skip to Content
Merck
All Photos(1)

Key Documents

BCR267

Indeno[1,2,3-cd]fluoranthene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
CHF 345.00

CHF 345.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
20 MG
CHF 345.00

About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein:
1970675
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

CHF 345.00


Estimated to ship on01 May 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc-2c(c1)-c3ccc4-c5ccccc5-c6ccc-2c3c46

InChI

1S/C22H12/c1-2-6-14-13(5-1)17-9-10-19-15-7-3-4-8-16(15)20-12-11-18(14)21(17)22(19)20/h1-12H

InChI key

XAKRHFYJDSMEMI-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR267

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service