Skip to Content
Merck
All Photos(1)

Key Documents

BCR172

Quartz (2.50 m2/g) (nitrogen BET specific surface area)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
CHF 219.00

CHF 219.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
CHF 219.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CHF 219.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

form

particles

manufacturer/tradename

JRC

application(s)

glass & ceramic

format

neat

Analysis Note

For more information please see:
BCR172

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service