Skip to Content
Merck
All Photos(1)

Key Documents

96556

Supelco

Tin(IV) sulfate – sulfuric acid solution

reagent, for the determination of nitrate acc. to DIN 38405-D9-3, suspension

Synonym(s):

Stannic sulfate in sulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn(SO4)2 · H2SO4
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

form

suspension

quality

for the determination of nitrate acc. to DIN 38405-D9-3

impurities

0.22-0.26% Sn
~80% H2SO4

density

1.74 g/mL at 20 °C

application(s)

general analytical

SMILES string

[Sn+4].OS(O)(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/3H2O4S.Sn/c3*1-5(2,3)4;/h3*(H2,1,2,3,4);/q;;;+4/p-4

InChI key

RAZPXVHCSOQXHL-UHFFFAOYSA-J

General description

precipitation has to be agitated

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service