93630
Trypsin, TPCK treated, from bovine pancreas
~7500 U/mg
Synonym(s):
TPCK-Trypsin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
specific activity
~7500 U/mg
shipped in
wet ice
storage temp.
−20°C
Unit Definition
1 U corresponds to the amount of enzyme which increases the absorbance at 253 nm by 0.001 per minute at pH 7.6 and 25°C (N-benzoyl-L-arginine ethyl ester, Cat. No. 12880, as substrate)
Other Notes
Sales restrictions may apply
TPCK-treatment irreversibly inhibits the chymotrypsin-activity without affecting the trypsin-activity; Application in (selective) hydrolysis/condensation of amide bonds; Tryptic degradation of opioid peptide precursors in rat brain
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Monatshefte fur Chemie / Chemical Monthly, 119, 1323-1323 (1988)
Chemical & pharmaceutical bulletin, 39(9), 2369-2372 (1991-09-01)
A high-performance liquid chromatographic method involving postcolumn fluorescence derivatization is described for the quantification of five fragment peptides (methionine-enkephalin-Thr-Ser-Glu-Lys, methionine-enkephalin-Lys, methionine-enkephalin-Arg, leucine-enkephalin-Lys and leucine-enkephalin-Arg) released by tryptic digestion from the opioid peptide precursors (proopiomeranocortin and proenkephalins A and B) in
Direct evidence for the presence of histidine in the active center of chymotrypsin.
Biochemistry, 2, 252-255 (1963-03-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service