Skip to Content
Merck
All Photos(1)

Key Documents

93572

Sigma-Aldrich

Formamide

puriss. p.a., ACS reagent, ≥99.5% (GC/T)

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
technique(s):
hybridization: suitable
bp:
210 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

Assay

≥99.5% (GC/T)

form

liquid

autoignition temp.

932 °F

expl. lim.

2.7-19 % (lit.)

technique(s)

hybridization: suitable

impurities

≤0.02% free acid (as HCOOH)
≤0.1% water

refractive index

n20/D 1.447 (lit.)
n20/D 1.447

pH

4-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

transition temp

solidification point 2-3 °C

density

1.134 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

suitability

complies for miscibility

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave spectrum and molecular structure of formamide.
Costain CC and Dowling JM.
J. Chem. Phys. , 32(1), 158-165 (1960)
J Casey et al.
Nucleic acids research, 4(5), 1539-1552 (1977-01-01)
The thermal stabilities of RNA:DNA hybrids are substantially greater than those of DNA:DNA duplexes in aqueous electrolyte solutions containing high concentrations of formamide. Association rates to form DNA:DNA duplexes and DNA:RNA hybrids have been measured in these solvents. There is
Solubilization in formamide protects RNA from degradation.
Chomczynski P.
Nucleic Acids Research, 20(14), 3791-3791 (1992)
The formamide method for the extraction of polysaccharides from haemolytic streptococci.
Fuller AT.
British Journal of Experimental Pathology, 32(1), 130-130 (1960)
Penelope A Bryant et al.
PloS one, 6(5), e19556-e19556 (2011-06-10)
A central issue in the design of microarray-based analysis of global gene expression is that variability resulting from experimental processes may obscure changes resulting from the effect being investigated. This study quantified the variability in gene expression at each level

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service