Skip to Content
Merck
All Photos(1)

Documents

92939

Supelco

4-Methylaminoantipyrine hydrochloride

analytical standard

Synonym(s):

1,2-Dihydro-1,5-dimethyl-4-(methylamino)-2-phenyl-3H-pyrazol-3-one monohydrochloride, N-Methylaminophenazone, Noramidopyrine hydrochloride, Noraminopyrine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O · HCl
CAS Number:
Molecular Weight:
253.73
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)
95.0-105.0% (AT)

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

O=C1C(NC)=C(C)N(C)N1C2=CC=CC=C2.[H]Cl

InChI

1S/C12H15N3O.ClH/c1-9-11(13-2)12(16)15(14(9)3)10-7-5-4-6-8-10;/h4-8,13H,1-3H3;1H

InChI key

NNONRTWOIVTAHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service