Skip to Content
Merck
All Photos(1)

Documents

90013

Supelco

Etofibrate

analytical standard

Synonym(s):

2-[(Pyridin-3-yl)carbonyloxy]ethyl 2-(4-chlorophenoxy)-2-methylpropanoate, 3-Pyridinecarboxylic acid 2-[2-(4-chlorophenoxy)-2-methyl-1-oxopropoxy]ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18ClNO5
CAS Number:
Molecular Weight:
363.79
Beilstein:
497931
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(OCCOC(C(C)(C)OC1=CC=C(Cl)C=C1)=O)C2=CC=CN=C2

InChI

1S/C18H18ClNO5/c1-18(2,25-15-7-5-14(19)6-8-15)17(22)24-11-10-23-16(21)13-4-3-9-20-12-13/h3-9,12H,10-11H2,1-2H3

InChI key

XXRVYAFBUDSLJX-UHFFFAOYSA-N

General description

Etofibrate (EF) is an ethylene glycol diester of clofibric and nicotinic acids. It is a cholesterol-lowering drug prescribed for treating hyperlipidaemias.

Application

Etofibrate may be used as an analytical reference standard for the determination of the analyte in plasma samples, pharmaceutical formulations, and environmental samples by analytical methods.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High performance liquid chromatographic determination of etofibrate and its hydrolysis products
El-Gindy A, et al.
Journal of Pharmaceutical and Biomedical Analysis, 43(1), 196-203 (2007)
Determination of 136 pharmaceuticals and hormones in sewage sludge using quick, easy, cheap, effective, rugged and safe extraction followed by analysis with liquid chromatography--time-of-flight-mass spectrometry
Peysson W and Vulliet E
Journal of Chromatography A, 1290(5), 46-61 (2013)
Pharmaceuticals in groundwaters: analytical methods and results of a monitoring program in Baden-W
Sacher F, et al.
Journal of Chromatography A, 938(1-2), 199-210 (2001)
Determination of etofibrate, fenofibrate, and atorvastatin in pharmaceutical preparations and plasma using differential pulse polarographic and square wave voltammetric techniques
Korany MA, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 91(5), 1051-1058 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service