Skip to Content
Merck
All Photos(1)

Key Documents

89600

Sigma-Aldrich

Methylhydroquinone

purum, ≥98.0% (HPLC)

Synonym(s):

Toluhydroquinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3-1,4-(OH)2
CAS Number:
Molecular Weight:
124.14
Beilstein:
2041489
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

autoignition temp.

851 °F

mp

126-128 °C
128-130 °C (lit.)

SMILES string

Cc1cc(O)ccc1O

InChI

1S/C7H8O2/c1-5-4-6(8)2-3-7(5)9/h2-4,8-9H,1H3

InChI key

CNHDIAIOKMXOLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

341.6 °F - closed cup

Flash Point(C)

172 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J W Priest et al.
Biochemistry, 28(23), 9192-9200 (1989-11-14)
A crude extract that catalyzes the epoxidation of toluquinol and gentisyl alcohol was isolated from cultures of Penicillium patulum. About 60% of the activity sedimented from crude extract upon centrifugation at 105,000g for 2 h, and at 30,000g for 30
Montira Leelakriangsak et al.
Molecular microbiology, 67(5), 1108-1124 (2008-01-23)
Recently, we showed that the MarR-type repressor YkvE (MhqR) regulates multiple dioxygenases/glyoxalases, oxidoreductases and the azoreductase encoding yvaB (azoR2) gene in response to thiol-specific stress conditions, such as diamide, catechol and 2-methylhydroquinone (MHQ). Here we report on the regulation of
Amparo Urios et al.
Free radical biology & medicine, 35(11), 1373-1381 (2003-12-04)
The induction of mutagenic and cytotoxic effects by simple phenolics, including catechol (CAT), 3,4-dihydroxyphenylacetic acid (DOPAC), hydroquinone (HQ), and 2,5-dihydroxyphenylacetic (homogentisic) acid (HGA), appears to occur through an oxidative mechanism based on the ability of these compounds to undergo autoxidation
C Furihata et al.
Japanese journal of cancer research : Gann, 84(3), 223-229 (1993-03-01)
The possible tumor-promoting and tumor-initiating activities of p-methylcatechol and methylhydroquinone in the pyloric mucosa of male F344 rats were studied. Ornithine decarboxylase (ODC) and replicative DNA synthesis (RDS) were used as markers of tumor promotion and DNA single strand scission
Melissa García-Caballero et al.
Biochemical pharmacology, 85(12), 1727-1740 (2013-04-23)
Toluquinol, a methylhydroquinone produced by a marine fungus, was selected in the course of a blind screening for new potential inhibitors of angiogenesis. In the present study we provide the first evidence that toluquinol is a new anti-angiogenic-compound. In a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service