Skip to Content
Merck
All Photos(1)

Key Documents

89467

Supelco

TISAB III solution

Synonym(s):

Total ionic strength adjustment buffer III solution

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
CHF 199.00

CHF 199.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
500 ML
CHF 199.00

About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

CHF 199.00


Estimated to ship on14 April 2025


Request a Bulk Order

form

liquid

Quality Level

density

1.21 g/mL at 20 °C

General description

Visit our Sensor Applications portal to learn more.

Application

for the fluoride determination in presence of Al(III)

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service