Skip to Content
Merck
All Photos(1)

Key Documents

89434

Supelco

Ceftriaxone disodium salt hemi(heptahydrate)

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N8Na2O7S3 · 3.5H2O
CAS Number:
Molecular Weight:
661.60
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

~10% water

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1C2SCC(CSC3=NC(=O)C(=O)N([Na])N3C)=C(N2C1=O)C([O-])=O)c4csc(N)n4.CO\N=C(/C(=O)N[C@H]5C6SCC(CSC7=NC(=O)C(=O)N([Na])N7C)=C(N6C5=O)C([O-])=O)c8csc(N)n8

InChI

1S/2C18H18N8O7S3.4Na.7H2O/c2*1-25-18(22-12(28)13(29)23-25)36-4-6-3-34-15-9(14(30)26(15)10(6)16(31)32)21-11(27)8(24-33-2)7-5-35-17(19)20-7;;;;;;;;;;;/h2*5,9,15H,3-4H2,1-2H3,(H5,19,20,21,23,27,29,31,32);;;;;7*1H2/q;;4*+1;;;;;;;/p-4/b2*24-8-;;;;;;;;;;;/t2*9-,15-;;;;;;;;;;;/m11.........../s1

InChI key

PMRZKYOXTPBIQF-MAODNAKNSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emphysematous pyelitis.
Kendall Luyt et al.
The Journal of pediatrics, 166(1), 201-201 (2014-10-01)
Ning Li et al.
Pediatrics, 133(4), e917-e922 (2014-03-26)
Our aim was to evaluate the clinical profile, treatment, and outcome of ceftriaxone-associated postrenal acute renal failure (PARF) in children. We retrospectively studied 31 consecutive cases from 2003 to 2012 for PARF after ceftriaxone treatment. There was no past history
Anthony R Flores et al.
The Journal of infectious diseases, 209(10), 1520-1523 (2013-12-07)
Invasive group A streptococcal (GAS) strains often have genetic differences compared to GAS strains from nonsterile sites. Invasive, "hypervirulent" GAS strains can arise from a noninvasive progenitor following subcutaneous inoculation in mice, but such emergence has been rarely characterized in
Yi Yao et al.
Pharmacoepidemiology and drug safety, 21(11), 1197-1201 (2012-07-05)
The incidence of adverse reactions to ceftriaxone, which is widely used in China, has gradually increased, with an associated increase in patient fatality. An analysis of the reported data from articles published in China highlights the importance and extent of
Michael S Northrop et al.
Journal of pediatric hematology/oncology, 37(1), e63-e66 (2014-06-01)
Ceftriaxone is a frequently used empiric antibiotic in children. Acute hemolysis is a rare side effect of ceftriaxone therapy associated with a high mortality rate. A 14-year-old boy suffering from Crohn disease developed bacterial pneumonia that was treated with ceftriaxone.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service