Skip to Content
Merck
All Photos(2)

Key Documents

89274

Supelco

(2S-cis)-(−)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 164.00

CHF 164.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
CHF 164.00

About This Item

Empirical Formula (Hill Notation):
C13H14N2O4
CAS Number:
Molecular Weight:
262.26
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

CHF 164.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]/D -9.0±1.5°, c = 0.8 in acetic acid

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

270-272 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)C[C@@H]1NC(=O)[C@H](Cc2ccccc2)NC1=O

InChI

1S/C13H14N2O4/c16-11(17)7-10-13(19)14-9(12(18)15-10)6-8-4-2-1-3-5-8/h1-5,9-10H,6-7H2,(H,14,19)(H,15,18)(H,16,17)/t9-,10-/m0/s1

InChI key

VNHJXYUDIBQDDX-UWVGGRQHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Degradation product from aspartame (E951)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timo Stark et al.
Journal of agricultural and food chemistry, 53(18), 7222-7231 (2005-09-01)
The taste compounds inducing the blood-like, metallic bitter taste sensation reported recently for a dichloromethane extract prepared from roasted cocoa nibs were identified as a series of 25 diketopiperazines by means of HPLC degustation, LC-MS/MS, and independent synthesis. Among these
S Kaakkola et al.
Brain research bulletin, 32(6), 667-672 (1993-01-01)
The effects of two diketopiperazines, Cyclo (His-Pro) (CHP) and Cyclo (Asp-Phe) (CAP), on striatal extracellular levels of dopamine (DA), dihydroxyphenylacetic acid (DOPAC), homovanillic acid (HVA), and 5-hydroxyindoleacetic acid (5-HIAA) were examined using in vivo microdialysis in anaesthetized rats. Treatment with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service