Skip to Content
Merck
All Photos(1)

Key Documents

80820

Sigma-Aldrich

Piperonal

purum, ≥99.0% (GC)

Synonym(s):

1,3-Benzodioxole-5-carboxaldehyde, 3,4-(Methylenedioxy)benzaldehyde, Heliotropin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O3
CAS Number:
Molecular Weight:
150.13
Beilstein:
131691
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

1 mmHg ( 87 °C)

grade

purum

Assay

≥99.0% (GC)

bp

264 °C (lit.)

mp

35-39 °C (lit.)
36-37 °C

solubility

methanol: 0.1 g/mL, clear

SMILES string

[H]C(=O)c1ccc2OCOc2c1

InChI

1S/C8H6O3/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-4H,5H2

InChI key

SATCULPHIDQDRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

150.1 °F

Flash Point(C)

65.62 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Dahl et al.
Biochemical pharmacology, 34(5), 631-636 (1985-03-01)
Eighteen methylenedioxyphenyl (MDP) compounds, including some commonly inhaled by people, were tested for the ability to inhibit rabbit nasal microsomal cytochrome P-450-dependent hexamethylphosphoramide (HMPA) N-demethylase. For comparison, liver microsomes were also used. Nasal cytochrome P-450 from rabbits metabolized MDP compounds
S Peock et al.
Journal of the Royal Society of Health, 113(6), 292-294 (1993-12-01)
Piperonal, once used to kill lice in Australian hospitals, was acclaimed as an effective pediculicide (Corlette, 1925) by the standards of the day. It is unusual in also exhibiting a repellent action against lice, a property only recently realised. A
Ariel Ceferino Toloza et al.
Memorias do Instituto Oswaldo Cruz, 101(1), 55-56 (2006-05-16)
New alternative insecticides are necessary for the chemical control of head lice. In this study the fumigant knockdown time 50% (KT50) and repellency index (RI) of three aliphatic lactones was compared with two essential oils and DDVP, against permethrin-resistance Pediculus
The inhibition of rat nasal cytochrome P-450-dependent mono-oxygenase by the essence heliotropin (piperonal).
A R Dahl
Drug metabolism and disposition: the biological fate of chemicals, 10(5), 553-554 (1982-09-01)
Patricia Guerra-Diaz et al.
Analytical chemistry, 82(7), 2826-2835 (2010-03-09)
A preconcentration device that targets the volatile chemical signatures associated with illicit drugs and explosives (high and low) has been designed to fit in the inlet of an ion mobility spectrometer (IMS). This is the first reporting of a fast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service