Skip to Content
Merck
All Photos(1)

Key Documents

75636

Sigma-Aldrich

Osmium tetroxide solution

2.5% in tert-butanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OsO4
CAS Number:
Molecular Weight:
254.23
MDL number:
UNSPSC Code:
12352300

contains

tert-butyl hydroperoxide as stabilizer

reaction suitability

reagent type: oxidant

concentration

2.5% in tert-butanol

density

0.81 g/mL at 20 °C
1.0 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

O=[Os](=O)(=O)=O

InChI

1S/4O.Os

InChI key

VUVGYHUDAICLFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service