Skip to Content
Merck
All Photos(1)

Key Documents

75360

Sigma-Aldrich

Tropaeolin 000 No. 1

for microscopy (Hist.), indicator (pH 7.6-8.9)

Synonym(s):

α-Naphthol orange, 4-(4-Hydroxy-1-naphthylazo)benzenesulfonic acid sodium salt, Orange I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N2NaO4S
CAS Number:
Molecular Weight:
350.32
Colour Index Number:
14600
Beilstein:
3826844
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

grade

for microscopy (Hist.)
indicator (pH 7.6-8.9)

Quality Level

form

crystals

technique(s)

titration: suitable

SMILES string

[Na+].Oc1ccc(\N=N\c2ccc(cc2)S([O-])(=O)=O)c3ccccc13

InChI

1S/C16H12N2O4S.Na/c19-16-10-9-15(13-3-1-2-4-14(13)16)18-17-11-5-7-12(8-6-11)23(20,21)22;/h1-10,19H,(H,20,21,22);/q;+1/p-1/b18-17+;

InChI key

AZLXCBPKSXFMET-ZAGWXBKKSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Staining of proteins[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I. Kalousek et al.
The International Journal of Biochemistry, 16, 121-121 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service