Skip to Content
Merck
All Photos(1)

Key Documents

68771

Supelco

Isoimperatorin

analytical standard

Synonym(s):

4-(3-Methylbut-2-enyloxy)furo[3,2-g]chromen-7-one, 4-[(3-Methyl-2-buten-1-yl)oxy]-7H-furo[3,2-g][1]benzopyran-7-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
Beilstein:
1291723
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O=C1C=CC(C(OCC=C(C)C)=C(C=CO2)C2=C3)=C3O1

InChI

1S/C16H14O4/c1-10(2)5-7-19-16-11-3-4-15(17)20-14(11)9-13-12(16)6-8-18-13/h3-6,8-9H,7H2,1-2H3

InChI key

IGWDEVSBEKYORK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng-jun Cai et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 31(8), 1160-1162 (2008-12-31)
To study chemical constituents of antibacterial activity fraction of Angelica polymorpha. Compounds were isolated by repeatedly silica gel column chromatography and recrystallization. Their structures were identified by physical and chemical evidences and spectral methods. Seven compounds were obtained from the
Hong Nie et al.
Chinese journal of integrative medicine, 15(6), 442-447 (2010-01-19)
To demonstrate the vasodilatation activity of the coumarin-containing Angelica dahurica var. formosana and to further analyze active components in the herb extracts. (1) The vasodilatation effects induced by different extracts (cyclohexane, ethyl acetate, acetone, methanol, 95 % ethanol and water)
Shinsuke Marumoto et al.
Bioorganic & medicinal chemistry, 18(1), 455-459 (2009-11-27)
Biotransformation studies conducted on the furanocoumarins isoimperatorin (1) and imperatorin (3) have revealed that 1 was metabolized by Glomerella cingulata to give the corresponding reduced acid, 6,7-furano-5-prenyloxy hydrocoumaric acid (2), and 3 was transformed by G. cingulata to give the
Ah Yeon Park et al.
Biomedical chromatography : BMC, 23(10), 1034-1043 (2009-04-30)
Rapid, simple and reliable HPLC/UV and LC-ESI-MS/MS methods for the simultaneous determination of five active coumarins of Angelicae dahuricae Radix, byakangelicol (1), oxypeucedanin (2), imperatorin (3), phellopterin (4) and isoimperatorin (5) were developed and validated. The separation condition for HPLC/UV
Jaroslaw Widelski et al.
Molecules (Basel, Switzerland), 14(8), 2729-2734 (2009-08-25)
The first phytochemical investigation of the fruits of Angelica lucida has led to the isolation and characterization of five known coumarins (imperatorin, isoimperatorin, heraclenol, oxypeucedanin hydrate and heraclenin). All isolated compounds were identified by means of spectral and literature data.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service