Skip to Content
Merck
All Photos(1)

Key Documents

64245

Sigma-Aldrich

tert-Butyl (1,2-methanofullerene C60)-61-carboxylate

purum, ≥95.0% (HPLC)

Synonym(s):

(1,2-Methanofullerene-C60)-61-carboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C60C(H)C00C(CH3)3
CAS Number:
Molecular Weight:
834.78
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

purum

Assay

≥95.0% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)C1C23c4c5c6c7c8c9c(c%10c%11c2c%12c4c%13c%14c5c%15c6c%16c8c%17c%18c9c%19c%10c%20c%11c%21c%12c%22c%13c%23c%14c%24c%15c%25c%16c%17c%26c%27c%18c%19c%28c%20c%29c%21c%22c%30c%23c%31c%24c%25c%26c%32c%27c%28c%29c%30c%31%32)C137

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for the synthesis of substituted fullerenes Interaction of C60 fullerence derivative with L-histidine was studied in detail. The quantum chemical calculations were focused on the enantioselective binding between the two.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Computational study of enantioselective interaction between C60 fullerene and its derivatives with L-histidine.
Lala B, et al.
Journal of Molecular Modeling, 13(4), 531-536 (2007)
L. Isaacs et al.
Helvetica Chimica Acta, 76, 1231-1231 (1993)
L. Isaacs et al.
Helvetica Chimica Acta, 76, 2454-2454 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service