Skip to Content
Merck
All Photos(1)

Documents

59659

Supelco

Toluidine Red

analytical standard

Synonym(s):

Pigment Red 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N3O3
CAS Number:
Molecular Weight:
307.30
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

270-272 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cc1ccc(\N=N\c2c(O)ccc3ccccc23)c(c1)[N+]([O-])=O

InChI

1S/C17H13N3O3/c1-11-6-8-14(15(10-11)20(22)23)18-19-17-13-5-3-2-4-12(13)7-9-16(17)21/h2-10,21H,1H3/b19-18+

InChI key

ZLFVRXUOSPRRKQ-VHEBQXMUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluidine Red is a synthetic organic dye belonging to the class of azo dyes, widely used for restoring or providing color to processed food products.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Toluidine Red may be used as a reference dye for the quantification of the analyte in food products using high-performance liquid chromatography with gel permeation chromatography clean-up procedure.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of fifteen illegal dyes in animal feeds and poultry products by ultra-high-performance liquid chromatography tandem mass spectrometry
Liu R, et al.
Journal of Chromatography A, 879(24), 2416-2422 (2011)
Ying Jiang et al.
Sexually transmitted diseases, 38(3), 244-245 (2010-09-28)
We retrospectively analyzed the Venereal Disease Research Laboratory test, Treponema pallidum particle agglutination test, and toluidine red unheated serum test (TRUST) in 41 cases of HIV-negative neurosyphilis and 34 non-neurologic syphilitic patient and found that serum-TRUST titers could be the
Simultaneous determination of 14 oil-soluble synthetic dyes in chilli products by high performance liquid chromatography with a gel permeation chromatography clean-up procedure
Zhu Y, et al.
Food Chemistry, 145(24), 956-962 (2014)
CI Pigment Red 3.
IARC monographs on the evaluation of carcinogenic risks to humans, 57, 259-267 (1993-01-01)
S A Larsen et al.
Journal of clinical microbiology, 17(2), 341-345 (1983-02-01)
Both the rapid plasma reagin (RPR) 18-mm circle card test and the toluidine red unheated serum test (TRUST) were designed to be more rapid than the Venereal Disease Research Laboratory slide test for the serodiagnosis of syphilis. The RPR card

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service