Skip to Content
Merck
All Photos(1)

Documents

573663

Supelco

Dowex G26 Ion Exchange Resin

hydrogen form, 25-30 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000
NACRES:
SB.52

product name

Dowex G26 hydrogen form, hydrogen form

Quality Level

form

beads

crosslinking

10 % cross-linked

parameter

150 °C max. temp.

moisture

45-52%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

diameter

22-25 mesh
600-700 μm

operating pH

0-14

capacity

2.0 meq/mL by wetted bed volume

density

1.22 g/mL (lit.)

bulk density

800 g/L (22.2 kg/cuft)

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Strongly acidic cation exchange resin for chemical processing and mining. This resin does NOT meet FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service