Skip to Content
Merck
All Photos(1)

Key Documents

56168

Supelco

Camphor benzalkonium methosulfate

analytical standard

Synonym(s):

4-[(2-Oxo-3-bornylidene)methyl]phenyltrimethylammonium methyl sulfate, N,N,N-Trimethyl-4-(2-oxoborn-3-ylidenemethyl)anilinum methyl sulfate, N,N,N-Trimethyl-4-[(4,7,7-trimethyl-3-oxobicyclo[2.2.1]hept-2-ylidene)methyl]benzenaminium methyl sulfate, BCM

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CHF 252.00

CHF 252.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
CHF 252.00

About This Item

Empirical Formula (Hill Notation):
C20H28NO · CH3SO4
CAS Number:
Molecular Weight:
409.54
EC Number:
UNSPSC Code:
12352212
NACRES:
NA.24

CHF 252.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤3.0% water

application(s)

environmental

format

neat

SMILES string

[O-]S(=O)(OC)=O.CC1(C)[C@]2([H])CC[C@@]1(C)C(/C2=C\C3=CC=C([N+](C)(C)C)C=C3)=O

InChI

1S/C20H28NO.CH4O3S/c1-19(2)17-11-12-20(19,3)18(22)16(17)13-14-7-9-15(10-8-14)21(4,5)6;1-5(2,3)4/h7-10,13,17H,11-12H2,1-6H3;1H3,(H,2,3,4)/q+1;/p-1/b16-13-;

InChI key

BPDGKTMMBDRVHQ-UNVLYCKESA-M

General description

Camphor benzalkonium methosulfate (CBM) is a highly photostable, fat-soluble organic ultraviolet (UV) filter approved for use in sunscreens/cosmetics to absorb UV rays. CBM is one of the most effective UV absorbers which offers protection against UVB radiations.[1][2]

Application

Camphor benzalkonium methosulfate may be used as a reference material for the determination of the analyte in sunscreen products by liquid-chromatographic method.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A reliable and environmentally-friendly liquid-chromatographic method for multi-class determination of fat-soluble UV filters in cosmetic products
Chisvert A, et al.
Analytica Chimica Acta, 790(10), 61-67 (2013)
The degradation products of UV filters in aqueous and chlorinated aqueous solutions
Santos A, et al.
Water Research, 46(10), 3167-3176 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service