Skip to Content
Merck
All Photos(1)

Key Documents

55587

Supelco

2-Heptanol

analytical standard

Synonym(s):

Methyl pentyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH3
CAS Number:
Molecular Weight:
116.20
Beilstein:
1719088
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.419-1.421
n20/D 1.420 (lit.)

bp

160-162 °C (lit.)

density

0.817 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCCCCC(C)O

InChI

1S/C7H16O/c1-3-4-5-6-7(2)8/h7-8H,3-6H2,1-2H3

InChI key

CETWDUZRCINIHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Heptanol is a saturated, chiral, aliphatic straight chain alcohol that belongs to the class of 2-alkanols. It is present as an odor component in cheese, fruits, food and beverages.

Application

2-Heptanol may be used as an analytical standard for the determination of the analyte in volatile compound mixtures, fruit samples and aqueous samples by various chromatography techniques.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F

Flash Point(C)

59 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hedwig Strohalm et al.
Journal of agricultural and food chemistry, 55(25), 10339-10344 (2007-11-17)
The enantiomeric compositions of the acetates, butanoates, hexanoates, and octanoates of the secondary alcohols 2-pentanol, 2-heptanol, and 2-nonanol were determined in yellow (Passiflora edulis f. flavicarpa) and purple (Passiflora edulis Sims) passion fruits. The compounds were isolated by means of
Sniffer to determine the odor of gas chromatographic effluents
Acree TE, et al.
Analytical Chemistry, 48(12), 1821-1822 (1976)
Christiane Geithe et al.
Chemical senses, 42(3), 181-193 (2016-12-06)
Key food odorants are the most relevant determinants by which we detect, recognize, and hedonically evaluate the aroma of foods and beverages. Odorants are detected by our chemical sense of olfaction, comprising a set of approximately 400 different odorant receptor
Densities, viscosities, and refractive indices of binary mixtures of acetophenone and 2-alkanols
Almasi M and Iloukhani H
Journal of Chemical and Engineering Data, 55(3), 1416-1420 (2010)
Flavour profiling of `Marion?and thornless blackberries by instrumental and sensory analysis
Du XF, et al.
Food Chemistry, 121(4), 1080-1088 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service