Skip to Content
Merck
All Photos(1)

Key Documents

52848

Supelco

Opipramol

analytical standard

Synonym(s):

4-[3-(5H-Dibenz[b,f]azepin-5-yl)-propyl]-1-piperazineethanol, NSC 169867

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H29N3O
CAS Number:
Molecular Weight:
363.50
Beilstein:
627076
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

OCCN(CC1)CCN1CCCN2C(C=CC=C3)=C3C=CC4=C2C=CC=C4

InChI

1S/C23H29N3O/c27-19-18-25-16-14-24(15-17-25)12-5-13-26-22-8-3-1-6-20(22)10-11-21-7-2-4-9-23(21)26/h1-4,6-11,27H,5,12-19H2

InChI key

YNZFUWZUGRBMHL-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nazli Esfahani-Bayerl et al.
Neuropsychologia, 81, 168-179 (2016-01-15)
The contributions of the hippocampal formation and adjacent regions of the medial temporal lobe (MTL) to memory are still a matter of debate. It is currently unclear, to what extent discrepancies between previous human lesion studies may have been caused
Benjamin Sauer et al.
Journal of pharmaceutical and biomedical analysis, 179, 113005-113005 (2019-12-10)
Energy-dispersive X-ray fluorescence spectrometry (EDXRF) is a suitable analytical procedure for screening drug products for Elemental Impurities (EI) according to ICH guideline Q3D. EDXRF represents a cost-efficient, robust and standard-free alternative compared to other methodologies for trace analysis, and therefore
Koleta Hemine et al.
Carbohydrate polymers, 250, 116957-116957 (2020-10-15)
It is widely believed that the hydrophobic effect governs the binding of guest molecules to cyclodextrins (CDs). However, it is also known that high hydrophobicity of guest molecules does not always translate to the formation of stable inclusion complexes with
Masoumeh Emamghoreishi et al.
Epilepsy research, 154, 74-76 (2019-05-12)
This study aimed to investigate the effects of sigma receptor modulators, opipramol and BD-1063, on epileptogenesis in pentylenetetrazole (PTZ)-kindling model of epilepsy. Mice (n = 6/group) were received PTZ (30 mg/kg), PTZ plus opipramol (5 or 10 mg/kg), PTZ plus opipramol (5 mg/kg) plus BD-1063
Lilian H J Richter et al.
Analytica chimica acta, 1070, 69-79 (2019-05-20)
Nonadherence to antihypertensive drugs therapy is known to be a serious issue in hypertension treatment. Liquid chromatography (LC) coupled to mass spectrometry (MS) was shown to allow the assessment of such nonadherence in blood and urine sample. However, their sampling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service