Skip to Content
Merck
All Photos(1)

Documents

52765

Sigma-Aldrich

Hexane

≥99.0% (GC), ACS reagent, puriss. p.a.

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

product name

Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

impurities

thiophene, in accordance
≤0.002% free acid (as CH3COOH)
≤0.005% sulfur compounds (as S)
≤0.05% water

evapn. residue

≤0.001%

refractive index

n20/D 1.375 (lit.)
n20/D 1.375

pH

7

bp

69 °C (lit.)

mp

−95 °C (lit.)

density

0.659 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
Sales restrictions may apply

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

G Di Nicola et al.
Journal of chromatography. A, 1190(1-2), 120-126 (2008-03-18)
Biodiesel (a mixture of fatty acid esters) is normally analyzed using gas chromatography/flame ionization detection, as specified by the ASTM D6584 and EN14105 standards. This paper proposes a binary gradient method for analyzing biodiesel mixtures using non-aqueous reverse phase HPLC
Comparative evaluation of 11 essential oils of different origin as functional antioxidants, antiradicals and antimicrobials in foods.
Sacchetti G, et al.
Food Chemistry, 91(4), 621-632 (2005)
Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)
Ali Mohraz et al.
Langmuir : the ACS journal of surfaces and colloids, 21(12), 5298-5306 (2005-06-01)
The development of model materials and image processing methods to directly visualize and quantify colloidal rod assembly by means of confocal laser scanning microscopy (CLSM) is reported. Monodisperse fluorescent colloidal rods are prepared by the uniaxial extensional deformation of sterically
Hilal E Toraman et al.
Journal of chromatography. A, 1359, 237-246 (2014-07-30)
The detailed compositional characterization of plastic waste pyrolysis oil was performed with comprehensive two-dimensional GC (GC×GC) coupled to four different detectors: a flame ionization detector (FID), a sulfur chemiluminescence detector (SCD), a nitrogen chemiluminescence detector (NCD) and a time of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service