Skip to Content
Merck
All Photos(1)

Key Documents

51881

Supelco

9-cis-Astaxanthin

analytical standard

Synonym(s):

(3S,3′S, 9-cis)-3,3′-Dihydroxy-β,β-carotene-4,4′-dione

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 1’010.00

CHF 1’010.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
CHF 1’010.00

About This Item

Empirical Formula (Hill Notation):
C40H52O4
CAS Number:
Molecular Weight:
596.84
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

CHF 1’010.00


Estimated to ship on25 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10% all-trans-Astaxanthin (HPLC)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−70°C

General description

9-cis-Astaxanthin is an isomer of astaxanthin (3,3 -dihydroxy-β,β-carotene-4,4 -dione), belonging to the xanthophyll family of oxygenated derivatives of carotenoids.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid baseline separation of enantiomers and a mesoform of all-trans-astaxanthin, 13-cis-astaxanthin, adonirubin, and adonixanthin in standards and commercial supplements.
Wang C, et al.
Journal of Chromatography A, 1194(2), 172-177 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service