ProClin 200 is an excellent biocide for in vitro diagnostic reagents that are sensitive to the high salt concentration of ProClin 150.
500402
ProClin™ 200
ProClin™ preservative for use in IVD manufacturing, CMIT/MIT 1.5%, pkg of 15 L
Synonym(s):
1.5% solution of 5-chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-4-isothiazolin-3-one
About This Item
Recommended Products
form
liquid
shelf life
18 mo.
contains
3% Mg and Cu salts
composition
CMIT/MIT, 1.5%
packaging
pkg of 15 L
parameter
operating temperature at <45 °C
matrix
Water
operating pH range
2.5-8.5
pH range
1.7-3.7
bp
100 °C
mp
-3 °C
solubility
water: soluble
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
When would I choose to use ProClin 200?
1 answer-
Helpful?
-
-
What is the concentration of active ingredients in ProClin 200?
1 answer-
ProClin 200 contains a total of 1.5% active ingredient.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
What is the matrix of the ProClin 200?
1 answer-
ProClin 200 is an aqueous solution, containing 0.15 - 0.17 % Cupric nitrate and 1.4 - 2.0 % Magnesium nitrate in water.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service