Skip to Content
Merck
All Photos(1)

Key Documents

487155

Supelco

Iodomethane-13C solution

99 atom % 13C (iodomethane-13C), NMR reference standard, 1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.2 %, trimethyl phosphite 1 %, NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CH3I
Molecular Weight:
142.93
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

Quality Level

isotopic purity

99 atom % 13C (iodomethane-13C)

composition

chromium(III) acetylacetonate, 0.2%
trimethyl phosphite, 1%

concentration

1% in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

suitability

suitable for NMR (reference standard)

application(s)

agriculture

format

single component solution

mass shift

M+1

SMILES string

[13CH3]I

InChI

1S/CH3I/c1-2/h1H3/i1+1

InChI key

INQOMBQAUSQDDS-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Indirect detection test

Quantity

3 mm O.D. tube contains 0.235 mL.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service