Skip to Content
Merck
All Photos(1)

Key Documents

47885

Sigma-Aldrich

Fumaraldehyde bis(dimethyl acetal)

technical, ≥90% (GC)

Synonym(s):

trans-1,1,4,4-Tetramethoxy-2-butene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CHCH=CHCH(OCH3)2
CAS Number:
Molecular Weight:
176.21
Beilstein:
1723349
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.428 (lit.)
n20/D 1.429

bp

100-103 °C/15 mmHg (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

COC(OC)\C=C\C(OC)OC

InChI

1S/C8H16O4/c1-9-7(10-2)5-6-8(11-3)12-4/h5-8H,1-4H3/b6-5+

InChI key

ZFGVCDSFRAMNMT-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Starting material for preparing the unstable mono-acetal, a versatile building block[1][2]; Synthesis of chiral cyclic N,O-acetals employed in asymmetric reactions[3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 44, 5929-5929 (1988)
G.M. Coppola
Synthesis, 1021-1021 (1984)
S. Cardani et al.
Tetrahedron, 44, 5563-5563 (1988)
R. Gree et al.
Tetrahedron Letters, 27, 4983-4983 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service