Skip to Content
Merck
All Photos(1)

Key Documents

47149

Sigma-Aldrich

5-Bromo-2-chloropyrimidine

purum, ≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2BrClN2
CAS Number:
Molecular Weight:
193.43
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (HPLC)

mp

73-79 °C (lit.)
74-77 °C

SMILES string

Clc1ncc(Br)cn1

InChI

1S/C4H2BrClN2/c5-3-1-7-4(6)8-2-3/h1-2H

InChI key

XPGIBDJXEVAVTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angeles Mosquera et al.
Organic letters, 10(17), 3745-3748 (2008-08-05)
The palladium-catalyzed cross-coupling reaction of triorganoindium reagents (R3In) with 5-bromo-2-chloropyrimidine proceeds chemoselectively, in good yields, to give 5-substituted-2-chloropyrimidines or 2,5-disubstituted pyrimidines using 40 or 100 mol % of R3In, respectively. Sequential cross-couplings are also performed, in one pot, using two
Gábor Vlád et al.
The Journal of organic chemistry, 67(18), 6550-6552 (2002-08-31)
A high-yield synthesis was developed for the preparation of 2,2'-bipyrimidine (1) using the Ullmann coupling of 2-iodopyrimidine. The new procedure was also used for the preparation of 4,4',6,6'-tetramethyl-2,2'-bipyrimidine (2) and 5,5'-dibromo-2,2'-bipyrimidine (3).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service