Skip to Content
Merck
All Photos(1)

Key Documents

46390

Supelco

Endrin-ketone solution

100 ng/μL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

3b,4,5,6,6,6a-Hexachlorodecahydro-2,5,7-metheno-3H-cyclopenta[a]pentalen-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl6O
CAS Number:
Molecular Weight:
380.91
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 ng/μL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

Cl[C@@H]1[C@@]2(Cl)[C@H]3[C@@H]4C[C@@H]5[C@H]3[C@](Cl)(C2(Cl)Cl)[C@@]1(Cl)[C@@H]5C4=O

InChI

1S/C12H8Cl6O/c13-8-9(14)6-2-1-3(7(6)19)4-5(2)11(9,16)12(17,18)10(4,8)15/h2-6,8H,1H2/t2-,3+,4+,5-,6+,8+,9-,10+,11-/m1/s1

InChI key

IZHZFAQWVKBTSL-YTMKBKNTSA-N

Application

Endrin-ketone solution may be used in the extraction of organochlorine pesticides from river water.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent microextraction of chlorinated pesticides.
De Jager LS and Andrews ARJ.
Chromatographia, 50(11-12), 733-738 (1999)
Chromous chloride reductions. V. Reaction of endrin ketone (II) with chromous chloride solution.
Chau ASY, et al.
Bulletin of Environmental Contamination and Toxicology, 6(6), 481-484 (1971)
R A Doong et al.
Journal of chromatography. A, 918(1), 177-188 (2001-06-14)
An analytical procedure was developed using headspace solid-phase microextraction (HS-SPME) for the determination of organochlorine pesticides (OCPs) and their metabolites in sandy soil samples. The developed procedures involving fiber selection, temperature effect, absorption time, soil matrix and the addition of
Ramandeep Kaur et al.
Molecules (Basel, Switzerland), 24(6) (2019-03-16)
Fabric phase sorptive extraction, an innovative integration of solid phase extraction and solid phase microextraction principles, has been combined with gas chromatography-mass spectrometry for the rapid extraction and determination of nineteen organochlorine pesticides in various fruit juices and water samples.
Archibold Buah-Kwofie et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1105, 85-92 (2018-12-24)
The monitoring of organic contaminants in the environment has gained tremendous traction in recent times, however, little attention has focused on the development of appropriate methods for the analysis of biological tissues from key sentinel species. In this study, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service