Skip to Content
Merck
All Photos(1)

Key Documents

46212

Supelco

3-Methyl-2-butanone

analytical standard

Synonym(s):

Isopropyl methyl ketone, Methyl isopropyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOCH3
CAS Number:
Molecular Weight:
86.13
Beilstein:
1071238
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

98-100%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.388 (lit.)

bp

94-95 °C (lit.)

mp

−92 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)C(C)=O

InChI

1S/C5H10O/c1-4(2)5(3)6/h4H,1-3H3

InChI key

SYBYTAAJFKOIEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F

Flash Point(C)

-1 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sami Sajjadifar et al.
Molecules (Basel, Switzerland), 15(4), 2491-2498 (2010-04-30)
Methyl indolenines (4a-c) and(5a-c) were prepared in high yield by a Fischer indole synthesis reaction of o,m-tolylhydrazine hydrochlorides (1a-b) with isopropyl methyl ketone (2) and 2-methylcyclohexanone (3) in acetic acid at room temperature. o,p- Nitrophenylhydrazines (1c-d) were reacted with 2-methylcyclohexanone
E M Gagan et al.
Archives of environmental contamination and toxicology, 52(3), 283-293 (2007-01-27)
Frequently the toxicity of an organic chemical mixture is close to dose-additive, even when the agents are thought to induce toxicity at different molecular sites of action. These findings appear to conflict with the hypothesis that a strictly dose-additive combined
Potential anticonvulsants IV: Condensation of isatin with benzoylacetone and isopropyl methyl ketone.
F D Popp et al.
Journal of pharmaceutical sciences, 71(9), 1052-1054 (1982-09-01)
S Schnell et al.
Biodegradation, 2(1), 33-41 (1991-01-01)
Strictly anaerobic bacteria were enriched and isolated from freshwater sediment sources in the presence and absence of sulfate with sorbic acid as sole source of carbon and energy. Strain WoSo1, a Gram-negative vibrioid sulfate-reducing bacterium which was assigned to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service