Skip to Content
Merck
All Photos(1)

Documents

46127

Sigma-Aldrich

7-Ethoxy-4-(trifluoromethyl)coumarin

suitable for fluorescence, ≥98.0% (TLC)

Synonym(s):

Ethyl 4-(trifluoromethyl)umbelliferyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9F3O3
CAS Number:
Molecular Weight:
258.19
Beilstein:
8555209
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥98.0% (TLC)

solubility

DMF: soluble
DMSO: soluble
methanol: soluble

fluorescence

λex 333 nm; λem 415 nm in methanol

suitability

suitable for fluorescence

SMILES string

CCOc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C12H9F3O3/c1-2-17-7-3-4-8-9(12(13,14)15)6-11(16)18-10(8)5-7/h3-6H,2H2,1H3

InChI key

OLHOIERZAZMHGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Substrate for monitoring the activity of cytochrome P450

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily E Scott et al.
Chemical research in toxicology, 15(11), 1407-1413 (2002-11-20)
Until recently, all known structures of bacterial cytochromes P450 suggested that substrate access to the buried active site occurred via the F-G region, a surface loop distal to the heme cavity. However, the structure of P450 51 indicates a large
Chitra Sridar et al.
The Journal of pharmacology and experimental therapeutics, 301(3), 945-952 (2002-05-23)
Tamoxifen is primarily used in the treatment of breast cancer. It has been approved as a chemopreventive agent for individuals at high risk for this disease. Tamoxifen is metabolized to a number of different products by cytochrome P450 enzymes. The
Ute M Kent et al.
Archives of biochemistry and biophysics, 423(2), 277-287 (2004-03-06)
Mechanistic studies with N-benzyl-1-aminobenzotriazole (BBT)-inactivated cytochrome P450 2B1 were conducted to determine which step(s) in the reaction cycle had been compromised. Stopped-flow studies, formation of the oxy-ferro intermediate, and analysis of products suggested that the reductive process was slower with
Shih-Feng Lan et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(4), 1314-1323 (2010-02-23)
In this study, we have evaluated the use of ultra-sterile alginate hydrogels encapsulated with HepG2 liver cells for applications in high throughput drug screening. We have studied the cellular viability and metabolic capacity of the encapsulated cells in two different
Rusty W Kelley et al.
Biochemistry, 44(7), 2632-2641 (2005-02-16)
The presence of one P450 can influence the catalytic characteristics of a second enzyme through the formation of heteromeric P450 complexes. Such a complex has been reported for mixed reconstituted systems containing NADPH-cytochrome P450 reductase, CYP2B4, and CYP1A2, where a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service