To convert a cation exhcange resin from the hydrogen form to the sodium form, use two to four bed volumes of 0.5-2.0% NaCl or NaOH.
44464
AmberChrom™ 50WX2
hydrogen form, strongly acidic, 200-400 mesh
About This Item
Recommended Products
description
cross-linkage 2%
Quality Level
form
beads
moisture
74-82%
technique(s)
LPLC: suitable
loss
~80% loss on drying, 110 °C
matrix
styrene divinylbenzene (gel)
matrix active group
sulfonic acid functional group
particle size
200-400 mesh
capacity
0.6 meq/mL by wetted bed volume
separation technique
cation exchange
InChI
1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2
InChI key
NWUYHJFMYQTDRP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How can I convert a cation exchange resin from the hydrogen form to the sodium form?
1 answer-
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service