Skip to Content
Merck
All Photos(1)

Key Documents

437530

Sigma-Aldrich

Trichloroethylene

≥99.5%, ACS reagent

Synonym(s):

TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade:
ACS reagent
Assay:
≥99.5%
bp:
86.7 °C (lit.)
86.7 °C
vapor pressure:
61 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

4.5 (vs air)

vapor pressure

61 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

770 °F

impurities

≤0.0001 meq/g Titr. Acid
≤0.0003 meq/g Titr. base
≤0.02% water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)
86.7 °C

mp

−84.8 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

cation traces

heavy metals: ≤1 ppm

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in poly-coated bottle

Other Notes

Contains free halogens

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda L Eiden et al.
Journal of medical entomology, 52(3), 429-436 (2015-09-04)
Permethrin is a commonly used acaricide for tick control on domestic animals and in residential environments, while fipronil use is restricted to on-animal treatment. Following widespread reports of permethrin and fipronil application failures to control indoor infestations of Rhipicephalus sanguineus
Jane C Caldwell et al.
Environmental health perspectives, 114(9), 1457-1463 (2006-09-13)
Trichloroethylene (TCE) exposure has been associated with increased risk of liver and kidney cancer in both laboratory animal and epidemiologic studies. The U.S. Environmental Protection Agency 2001 draft TCE risk assessment concluded that it is difficult to determine which TCE
Glinda S Cooper et al.
Environmental health perspectives, 117(5), 696-702 (2009-05-30)
Our objective was to examine experimental and epidemiologic studies pertaining to immune-related, and specifically autoimmune-related, effects of trichloroethylene (TCE). We performed a literature search of PubMed and reviewed bibliographies in identified articles. We then systematically reviewed immune-related data, focusing on
Michihiro Kamijima et al.
International archives of occupational and environmental health, 80(5), 357-370 (2006-11-16)
Workers exposed to trichloroethylene (TCE) rarely show severe generalized skin disorders and accompanying hepatitis which resemble drug hypersensitivities. The disorders are completely different from solvent-induced irritating contact dermatitis, and their serious consequences have become one of the critical occupational health
Cheryl Siegel Scott et al.
Environmental health perspectives, 114(9), 1471-1478 (2006-09-13)
A large body of epidemiologic evidence exists for exploring causal associations between cancer and trichloroethylene (TCE) exposure. The U.S. Environmental Protection Agency 2001 draft TCE health risk assessment concluded that epidemiologic studies, on the whole, support associations between TCE exposure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service