Skip to Content
Merck
All Photos(1)

Key Documents

41924

Supelco

Cyanoguanidine

pharmaceutical impurity standard, ≥95.0% (HPLC)

Synonym(s):

Dicyandiamide, Metformin Related Compound A (USP), Metformin impurity A (PhEur), Cyanoguanidine, Dicyanodiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NHCN
CAS Number:
Molecular Weight:
84.08
Beilstein:
605637
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% related substances
≤5.0% solvents
≤5.0% water

mp

208-211 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=N)NC#N

InChI

1S/C2H4N4/c3-1-6-2(4)5/h(H4,4,5,6)

InChI key

QGBSISYHAICWAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Li-Mei Zhang et al.
The ISME journal, 6(5), 1032-1045 (2011-12-03)
Increasing evidence demonstrated the involvement of ammonia-oxidizing archaea (AOA) in the global nitrogen cycle, but the relative contributions of AOA and ammonia-oxidizing bacteria (AOB) to ammonia oxidation are still in debate. Previous studies suggest that AOA would be more adapted
F Al-Rimawi
Talanta, 79(5), 1368-1371 (2009-07-29)
A simple, and stability-indicating liquid chromatographic method was developed and validated for the analysis of metformin hydrochloride and its related compound (1-cyanoguanidine) in tablet formulations. Liquid chromatography with a UV detector at a wavelength of 232 nm using a Nova-Pak
Enrico Colli et al.
Expert opinion on investigational drugs, 16(7), 999-1007 (2007-06-28)
'Overactive bladder' (OAB) is a syndrome that is characterised by symptoms of urgency, with or without urge urinary incontinence, usually with frequency and nocturia [1] . It is a highly prevalent condition affecting 17% of the general population, with a
Xianglan Li et al.
Chemosphere, 75(10), 1417-1422 (2009-03-10)
A field experiment was conducted to study the effect of timing of joint application of urease inhibitor hydroquinone (HQ) and nitrification inhibitor dicyandiamide (DCD) on N(2)O emission from irrigated lowland rice paddy field. Four treatments including Treatment CK (the control
Hong Nie et al.
Bioorganic & medicinal chemistry letters, 16(21), 5513-5516 (2006-08-29)
A series of N-(2-hydroxy-3-sulfonamidobenzene)-N'-arylcyanoguanidines was prepared. In general, these compounds proved to be potent antagonists of CXCR2 while the selectivity versus CXCR1 ranged from non-selective to >200-fold.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service