Skip to Content
Merck
All Photos(1)

Key Documents

41663

Sigma-Aldrich

1,3-Dimethyl-1,1,3,3-tetraphenyldisilazane

≥98.0% (NT)

Synonym(s):

1,1,3,3-Tetraphenyl-1,3-dimethyldisilazane, TPDMDS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3Si(C6H5)2]2NH
CAS Number:
Molecular Weight:
409.67
Beilstein:
2952534
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

form

solid

mp

87-89 °C (lit.)
87-89 °C

SMILES string

C[Si](N[Si](C)(c1ccccc1)c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C26H27NSi2/c1-28(23-15-7-3-8-16-23,24-17-9-4-10-18-24)27-29(2,25-19-11-5-12-20-25)26-21-13-6-14-22-26/h3-22,27H,1-2H3

InChI key

YFONAHAKNVIHPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dimethyl-1,1,3,3-tetraphenyldisilazane is a silylating reagent, used for the hydrophobization of glass or silica surfaces.[1]

Other Notes

For the deactivation of glass capillary columns by persilylation[2][2][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher A Girkin et al.
Investigative ophthalmology & visual science, 58(9), 3759-3769 (2017-07-25)
This study quantified the thickness and depth of the lamina cribrosa (LC) and peripapillary scleral thickness in high-resolution three-dimensional (3D) fluorescent reconstructions of the optic nerve head (ONH) in eyes from donors of African (AD) and European descent (ED). A
Silicon in Polymer Synthesis (2012)
Concentration modulation by thermal decomposition for multiplex gas chromatography.
J R Valentin et al.
HRC & CC. Journal of high resolution chromatography & chromatography communications, 6, 621-622 (1983-11-01)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service