Skip to Content
Merck
All Photos(1)

Key Documents

41625

Sigma-Aldrich

Dimethyl sulfide

≥95.0% (GC)

Synonym(s):

DMS, Methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S
CAS Number:
Molecular Weight:
62.13
Beilstein:
1696847
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

vapor density

2.1 (vs air)

vapor pressure

26.24 psi ( 55 °C)
7.79 psi ( 20 °C)

Assay

≥95.0% (GC)

autoignition temp.

402 °F

expl. lim.

19.7 %

refractive index

n20/D 1.435 (lit.)
n20/D 1.435

bp

38 °C (lit.)

mp

−98 °C (lit.)

solubility

H2O: soluble 7.28 g/L at 20 °C

density

0.846 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CSC

InChI

1S/C2H6S/c1-3-2/h1-2H3

InChI key

QMMFVYPAHWMCMS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfide (DMS) is formed in sea water due to various biological processes. It is the major sulfur containing gas generated by ocean and liberated to the atmosphere. Its quantitative determination during DMS generation in seawater tanks during phytoplankton bloom simulations has been reported.[1] Mechanism of its photochemical oxidation in atmosphere has been described.[2]

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-32.8 °F - closed cup

Flash Point(C)

-36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimethyl sulfide production during natural phytoplanktonic blooms.
Nguyen BC, et al.
Marine Chemistry, 24(2), 133-141 (1988)
B P Lomans et al.
Applied and environmental microbiology, 65(8), 3641-3650 (1999-07-31)
A newly isolated methanogen, strain DMS1(T), is the first obligately anaerobic archaeon which was directly enriched and isolated from a freshwater sediment in defined minimal medium containing dimethyl sulfide (DMS) as the sole carbon and energy source. The use of
Yin F, et al.
Journal of Atmospheric Chemistry, 11(4), 309-364 (1990)
Sheree Yau et al.
The ISME journal, 7(10), 1944-1961 (2013-04-27)
Organic Lake is a shallow, marine-derived hypersaline lake in the Vestfold Hills, Antarctica that has the highest reported concentration of dimethylsulfide (DMS) in a natural body of water. To determine the composition and functional potential of the microbial community and
Hayley E Arnold et al.
Global change biology, 19(4), 1007-1016 (2013-03-19)
The production of the marine trace gas dimethyl sulfide (DMS) provides 90% of the marine biogenic sulfur in the atmosphere where it affects cloud formation and climate. The effects of increasing anthropogenic CO2 and the resulting warming and ocean acidification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service