Skip to Content
Merck
All Photos(1)

Documents

41506

Sigma-Aldrich

Lactophenol-picric acid solution

for microscopy

Synonym(s):

Picric acid-lactophenol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
MA.02

grade

for microscopy

Quality Level

product line

BioChemika

form

liquid

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

application(s)

food and beverages
hematology
histology

storage temp.

room temp

suitability

fungi

General description

Picric acid (2,4,6-trinitrophenol) has antibacterial as well as antifungal properties. It stains the fungal spores and hyphae a pale to dark yellow color.

Components

Composition:
Phenol 100 g, Lactic acid 100 mL,
Glycerol 200 mL, Picric acid (saturated aqueous solution) 100 ml

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yiqun Mo et al.
Nanotoxicology, 14(9), 1175-1197 (2020-09-15)
We and other groups have demonstrated that exposure to nickel nanoparticles (Nano-Ni) results in severe and persistent lung inflammation and fibrosis, but the underlying mechanisms remain unclear. Here, we propose that miR-21 may play an important role in Nano-Ni-induced lung

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service