Skip to Content
Merck
All Photos(1)

Documents

41406

Supelco

4-Phenyl-2-butanone

analytical standard

Synonym(s):

Benzylacetone, Methyl 2-phenethyl ketone, NSC 44829, NSC 813

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2COCH3
CAS Number:
Molecular Weight:
148.20
Beilstein:
1907123
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.512 (lit.)

bp

235 °C (lit.)

density

0.989 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(=O)CCc1ccccc1

InChI

1S/C10H12O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3

InChI key

AKGGYBADQZYZPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Phenyl-2-butanone, a volatile aromatic ketone, is a fragrance compound present as a ubiquitous attractant in flowers.

Application

4-Phenyl-2-butanone may be used as an analytical standard for the determination of the analyte in volatile oils, as well as agarwood extracts of Aquilaria malaccensis, yeast cell culture, and biological samples by various chromatography based techniques.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kitamura et al.
Drug metabolism and disposition: the biological fate of chemicals, 27(7), 767-769 (1999-06-29)
The reductive metabolism in vivo of a flavoring additive, trans-4-phenyl-3-buten-2-one (PBO; trans-methyl styryl ketone) was investigated in rats and dogs. In both species, the double bond-reduced product, 4-phenyl-2-butanone (PBA), was detected by HPLC as the predominant species in blood after
Danny Kessler et al.
The Plant journal : for cell and molecular biology, 49(5), 840-854 (2007-02-24)
Flowers produce a plethora of secondary metabolites but only nectar sugars, floral pigments and headspace volatiles have been examined in the context of pollinator behavior. We identify secondary metabolites in the headspace and nectar of glasshouse- and field-grown Nicotiana attenuata
Takashi Miyoshi et al.
Biological & pharmaceutical bulletin, 36(9), 1474-1481 (2013-09-03)
Benzylacetone is released by heated agarwood, when inhaled it has a potent effect on reducing the locomotor activity of mice. This study investigated the relationships between the sedative activities of benzylacetone and its derivatives as well as the chemical structures
Patterns and consequences of benzyl acetone floral emissions from Nicotiana attenuata plants
Baldwin IT, et al.
Journal of Chemical Ecology, 23(10), 2327-2343 (1997)
Gas chromatography-mass spectrometry analysis of agarwood extracts from mature and juvenile Aquilaria malaccensis
Tsan PLJP and Mohamed R
International Journal of Agriculture and Biology, 16(3) (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service