Skip to Content
Merck
All Photos(2)

Key Documents

41373

Supelco

Heptenophos

PESTANAL®, analytical standard

Synonym(s):

7-Chlorobicyclo[3.2.0]hepta-2,6-dien-6-yl dimethyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12ClO4P
CAS Number:
Molecular Weight:
250.62
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 42.27-43.99%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

-10 to -25°C

SMILES string

COP(=O)(OC)OC1=C(Cl)C2C=CCC12

InChI

1S/C9H12ClO4P/c1-12-15(11,13-2)14-9-7-5-3-4-6(7)8(9)10/h3-4,6-7H,5H2,1-2H3

InChI key

GBAWQJNHVWMTLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Distribution of chostaquick in the rat body after different ways of intake].
D B Girenko et al.
Gigiena i sanitariia, (1)(1), 57-59 (1990-01-01)
Piia Jõul et al.
Analytical methods : advancing methods and applications, 13(1), 69-76 (2020-12-09)
The current study is focused on the in situ synthesis of a carbon aerogel (CA)-based solid-phase microextraction (SPME) fiber coating on stainless steel wire and evaluation of the suitability of CAs as SPME coating materials for the analysis of selected
Mario Masís-Mora et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1156, 122296-122296 (2020-08-24)
Biopurification systems are useful in the management of pesticide residues and provide an option to dispose wastewaters of agricultural origin derived from pesticide application practices. The analysis of pesticide residues in the biopurification system biomixture is necessary to determine whether
Mario Masís-Mora et al.
Chemosphere, 233, 733-743 (2019-06-15)
Biopurification systems (BPS) have been barely explored for removing complex mixtures of pesticides. In this study, the potential of a biomixture to remove simultaneously a mixture of herbicides (triazines), fungicides (triazoles) and insecticides (organophosphates) is presented. Also, a BPS using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service