Skip to Content
Merck
All Photos(1)

Key Documents

39406

Millipore

Mannitol Fermentation Membrane

white colored, sterile, diameter 70 mm, suitable for microbiology

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
CHF 24.50

CHF 24.50


Estimated to ship on28 May 2025


Request more information

Select a Size

Change View
25 DISCS
CHF 24.50

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

CHF 24.50


Estimated to ship on28 May 2025


Request more information

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
food and beverages
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Citrobacter spp.
Enterobacter spp.
Escherichia coli
Klebsiella spp.
Salmonella spp.
Serratia spp.
Shigella spp.
bacteria

General description

Mannitol Fermentation Membranes are substrate-carrying ID membranes used for rapid identification and differentiation of mannitol fermenting microorganisms from mixed flora. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The principle is based on the acid production during the fermentation, as indicated by the color change of a pH indicator to yellow.

Application

Mannitol Fermentation ID Membrane is used for rapid detection & confirmation of mannitol fermenting organisms in water and food samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostics, etc.

Analysis Note

Organism that can ferment Mannitol Citrobacter freundii (8090)Enterobacter aerogenes (13048)Escherichia coli (25922)Klebsiella pneumoniae (13883) Salmonella typhimurium (14028)Salmonella typhi (6539)Serratia marcescens (8100)Shigella flexneri (12022)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service