Skip to Content
Merck
All Photos(1)

Documents

37476

Supelco

AQUANAL-plus tin (Sn) 0.1-1.2 mg/L

refill pack for 37436

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41113319

application(s)

environmental
general analytical

Looking for similar products? Visit Product Comparison Guide

General description

Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations.

Analysis Note

also for photometric evaluation

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service