Skip to Content
Merck
All Photos(1)

Documents

37120

rac-1,2-Diheptadecanoylglycerol

≥99.0% (TLC)

Synonym(s):

D,L-α,β-Dimargarin, rac.-1,2-Dimargarinoyl-glycerol, rac.-Glycerol 1,2-dimargarinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H72O5
CAS Number:
Molecular Weight:
596.96
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥99.0% (TLC)

form

solid

mp

65-67 °C

SMILES string

CCCCCCCCCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCCCCCCCCCCC

InChI

1S/C37H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-36(39)41-34-35(33-38)42-37(40)32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35,38H,3-34H2,1-2H3

InChI key

RGROBSGNJZWWRZ-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Ogawa et al.
Rapid communications in mass spectrometry : RCM, 31(11), 928-936 (2017-04-04)
The acylglycerols in miso have not been studied although it is known that they are important to the taste. In order to determine the fatty acid constituents in the acylglycerols and analyze them individually, multiple reaction monitoring (MRM) was performed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service