Skip to Content
Merck
All Photos(2)

Key Documents

37005

Supelco

Oxaziclomefone

PESTANAL®, analytical standard

Synonym(s):

3-[1-(3,5-Dichlorophenyl)-1-methylethyl]-3,4-dihydro-6-methyl-5-phenyl-2H-1,3-oxazin-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 363.00

CHF 363.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
CHF 363.00

About This Item

Empirical Formula (Hill Notation):
C20H19Cl2NO2
CAS Number:
Molecular Weight:
376.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

CHF 363.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(C(=O)N(CO1)C(C)(C)c2cc(Cl)cc(Cl)c2)c3ccccc3

InChI

1S/C20H19Cl2NO2/c1-13-18(14-7-5-4-6-8-14)19(24)23(12-25-13)20(2,3)15-9-16(21)11-17(22)10-15/h4-11H,12H2,1-3H3

InChI key

FCOHEOSCARXMMS-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service