Skip to Content
Merck
All Photos(1)

Key Documents

36514

Supelco

Terbumeton-desethyl

PESTANAL®, analytical standard

Synonym(s):

2-Amino-4-tert-butylamino-6-methoxy-1,3,5-triazine, Desethylterbumetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N5O
CAS Number:
Molecular Weight:
197.24
Beilstein:
516008
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

NC1=NC(OC)=NC(NC(C)(C)C)=N1

InChI

1S/C8H15N5O/c1-4-5(2)10-7-11-6(9)12-8(13-7)14-3/h5H,4H2,1-3H3,(H3,9,10,11,12,13)

InChI key

FUNDQXLDYFURSA-UHFFFAOYSA-N

General description

Terbumeton-desethyl is a major degradation product of terbumeton, a methoxytriazine herbicide, which is found to be a persistent environmental contaminant.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Terbumeton-desethyl may be used as an analytical reference standard for the quantification of the analyte in honey, honey bees,[2] fruits and vegetables[3] using various chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficiency of QuEChERS approach for determining 52 pesticide residues in honey and honey bees.
Calatayud-Vernich P, et al.
MethodsX, 3, 452-458 (2016)
Application of gas chromatography-(triple quadrupole) mass spectrometry with atmospheric pressure chemical ionization for the determination of multiclass pesticides in fruits and vegetables.
Cherta L, et al.
Journal of Chromatography A, 1314, 224-240 (2013)
Leaching of terbumeton and terbumeton-desethyl from mini-columns packed with soil aggregates in laboratory conditions.
Conrad A, et al.
Chemosphere, 65(9), 1600-1609 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service